Learn More
Biolegend APC/Cyanine7 anti-human CD117
Catalog No. 502722400
Encompass_Preferred
APC/Cyanine7 anti-human CD117 (c-kit) [104D2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2400 Supplier Biolegend Supplier No. 313227
APC/Cyanine7 anti-human CD117 (c-kit) [104D2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests