Learn More
Biolegend APC/Cyanine7 anti-human CD235a
Supplier:  Biolegend 379007
Encompass_Preferred
APC/Cyanine7 anti-human CD235a (Glycophorin A) Recombinant [QA19A08] Isotype Mouse IgG2a Reactivity Human Apps FC Size 25 tests
Catalog No. 50-310-8018