Learn More
Biolegend APC/Cyanine7 anti-human CD314 (NKG2D)
Supplier:  Biolegend 320824
Encompass_Preferred
APC/Cyanine7 anti-human CD314 (NKG2D) [1D11]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1630781