Learn More
Biolegend APC/Cyanine7 anti-mouse CD122 (IL-2Rβ)
Supplier:  Biolegend 123221
Encompass_Preferred
APC/Cyanine7 anti-mouse CD122 (IL-2Rβ) [TM-β1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0497