Learn More
Biolegend APC/CYANINE7 ANTIHUM CD223 25T
Supplier:  Biolegend 369347
Encompass_Preferred
APC/Cyanine7 anti-human CD223 (LAG-3) Antibody25 testsCD223, also known as LAG-3, is a 70 kD type I transmembrane glycoprotein that is involved in T-cell signaling. Similar to CD4, CD223 binds MHC class II, but with a higher affinity. CD223 negatively regulates T-cell activation. It is expressed by activated T-cells and natural killer cells (NKs), as well as regulatory T-cells. It is transiently expressed on the surface of activated T-cells in acute conditions but high expression is maintained under tolerizing conditions. CD223 deficiency results in reduced tumor growth. CD223 and PD-1 can act in synergy and reverse exhausted phenotypes, improve tumor rejection, and control viral load.
Catalog No. NC2293350