Learn More
ABclonal Technology APC/Cyanine7 Rabbit anti- IgM
SDP
Catalog No. 5000013783
Encompass_Preferred
Enables several functions including antigen binding activity identical protein binding activity and immunoglobulin receptor binding activity Acts upstream of or within several processes including early endosome to late endosome transport immunoglobulin mediated immune response and positive regulation of B cell proliferation Located in external side of plasma membrane extracellular space and perinuclear region of cytoplasm Part of B cell receptor complex and immunoglobulin complex circulating Is expressed in several structures including brain hemolymphoid system and liver Used to study non-Hodgkin lymphoma and type 1 diabetes mellitus Human ortholog(s) of this gene implicated in agammaglobulinemia Orthologous to human IGHM (immunoglobulin heavy constant mu)
Catalog No. 50-000-13783 Supplier ABclonal Technology Supplier No. AS176100T
Enables several functions including antigen binding activity identical protein binding activity and immunoglobulin receptor binding activity Acts upstream of or within several processes including early endosome to late endosome transport immunoglobulin mediated immune response and positive regulation of B cell proliferation Located in external side of plasma membrane extracellular space and perinuclear region of cytoplasm Part of B cell receptor complex and immunoglobulin complex circulating Is expressed in several structures including brain hemolymphoid system and liver Used to study non-Hodgkin lymphoma and type 1 diabetes mellitus Human ortholog(s) of this gene implicated in agammaglobulinemia Orthologous to human IGHM (immunoglobulin heavy constant mu)