Learn More
Custom Networks Inc APC External Battery Pack, 3U, Li-Ion, 16 Ah, Rackmount
SDP
Catalog No. NC3978293
Encompass_Preferred
This external battery pack is designed to enhance the runtime of Smart-UPS Modular Ultra models. It comes with an external battery pack frame and four genuine battery modules, offering a reliable solution for increased power autonomy.
  • Compatible with all Smart-UPS Modular Ultra models.
  • Utilizes lithium-ion battery modules.
  • Allows adding additional battery modules to a Smart-UPS Modular UPS or external battery chassis to increase runtime.
  • Standard warranty of 5 years repair or replace.
  • Includes external battery pack frame and 4 genuine battery modules.
  • Battery is removable.
  • User replaceable.
Catalog No. NC3978293 Supplier Custom Networks Inc Supplier No. SRYLRMXBP
This external battery pack is designed to enhance the runtime of Smart-UPS Modular Ultra models. It comes with an external battery pack frame and four genuine battery modules, offering a reliable solution for increased power autonomy.
  • Compatible with all Smart-UPS Modular Ultra models.
  • Utilizes lithium-ion battery modules.
  • Allows adding additional battery modules to a Smart-UPS Modular UPS or external battery chassis to increase runtime.
  • Standard warranty of 5 years repair or replace.
  • Includes external battery pack frame and 4 genuine battery modules.
  • Battery is removable.
  • User replaceable.