Learn More
Jackson Immuno Research Labs Allophycocyanin (APC) AffiniPure F(ab')₂ Fragment Goat Anti-Mouse IgG (H+L), .5ml
SDP
Supplier:  Jackson Immuno Research Labs 115136146
Encompass_Preferred
Allophycocyanin-AffiniPure F(ab')2 Fragment Goat Anti-Mouse IgG (H+L) (min X Hu,Bov,Hrs,Rb,Sw Sr Prot)
Catalog No. NC9256961