Learn More
Biolegend APC/Fire™ 750 anti-mouse CD49b (pan-NK cells)
Supplier:  Biolegend 108925
Encompass_Preferred
APC/Fire™ 750 anti-mouse CD49b (pan-NK cells) [DX5]; Isotype: Rat IgM, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0071