Learn More
Biolegend APC/Fire™ 810 anti-mouse TER-119/Erythroid Cells
Supplier:  Biolegend 116263
Encompass_Preferred
APC/Fire™ 810 anti-mouse TER-119/Erythroid Cells [TER-119]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0273