Learn More
Biolegend APC/Fire™ 750 anti-human CD25
Supplier:  Biolegend 302642
Encompass_Preferred
APC/Fire™ 750 anti-human CD25 [BC96]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1900961