Learn More
Biolegend APC/Fire™ 750 anti-mouse Ly-6C
Supplier:  Biolegend 128046
Encompass_Preferred
APC/Fire™ 750 anti-mouse Ly-6C [HK1.4]; Isotype: Rat IgG2c, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-112-9841