Learn More
Biolegend APC/Fire™ 750 anti-H2A.X Phospho (Ser139)
Supplier:  Biolegend 613421
Encompass_Preferred
APC/Fire™ 750 anti-H2A.X Phospho (Ser139) [2F3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-208-6724