Learn More
Biolegend APC/Fire™ 750 anti-human CD115 (CSF-1R)
Supplier:  Biolegend 347324
Encompass_Preferred
APC/Fire™ 750 anti-human CD115 (CSF-1R) [9-4D2-1E4]; Isotype: Rat IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-207-6829