Learn More
Biolegend APC/Fire™ 750 anti-human CD123
Supplier:  Biolegend 396715
Encompass_Preferred
APC/Fire™ 750 anti-human CD123 [S18016F]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-9250