Learn More
Biolegend APC/Fire™ 750 anti-human CD158 (KIR2DL1/S1/S3/S5)
Supplier:  Biolegend 339520
Encompass_Preferred
APC/Fire™ 750 anti-human CD158 (KIR2DL1/S1/S3/S5) [HP-MA4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Cynomolgus; Apps: FC; Size: 100 tests
Catalog No. 50-207-6405