Learn More
Biolegend APC/Fire™ 750 anti-human CD162
Supplier:  Biolegend 328814
Encompass_Preferred
APC/Fire™ 750 anti-human CD162 [KPL-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1711119