Learn More
Biolegend APC/Fire™ 750 anti-human CD134 (OX40)
Supplier:  Biolegend 350032
Encompass_Preferred
APC/Fire™ 750 anti-human CD134 (OX40) [Ber-ACT35 (ACT35)]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. NC1320612