Learn More
Biolegend APC/Fire™ 750 anti-human CD19
Supplier:  Biolegend 363030
Encompass_Preferred
APC/Fire™ 750 anti-human CD19 [SJ25C1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1240564