Learn More
Biolegend APC/Fire™ 750 anti-human CD40
Supplier:  Biolegend 334345
Encompass_Preferred
APC/Fire™ 750 anti-human CD40 [5C3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1551178