Learn More
Biolegend APC/Fire™ 750 anti-human CD45RA [HI100]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 500 uL
Supplier:  Biolegend 983010
Encompass_Preferred
APC/Fire™ 750 anti-human CD45RA [HI100]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 500 uL
Catalog No. 50-237-7003