Learn More
Biolegend APC/Fire™ 750 anti-mouse/human CD11b
Supplier:  Biolegend 101262
Encompass_Preferred
APC/Fire™ 750 anti-mouse/human CD11b [M1/70]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Chimpanzee, Rabbit (Lapine); Apps: FC; Size: 100 ug
Catalog No. NC1084665