Learn More
Biolegend APC/Fire™ 750 anti-mouse CD11c
Supplier:  Biolegend 117352
Encompass_Preferred
APC/Fire™ 750 anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1552915