Learn More
Biolegend APC/Fire™ 750 anti-mouse CD45
Supplier:  Biolegend 103154
Encompass_Preferred
APC/Fire™ 750 anti-mouse CD45 [30-F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1361524