Learn More
Biolegend APC/Fire™ 750 anti-mouse I-A/I-E
Supplier:  Biolegend 107652
Encompass_Preferred
APC/Fire™ 750 anti-mouse I-A/I-E [M5/114.15.2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1499932