Learn More
Biolegend APC/Fire™ 750 anti-mouse CD49d
Supplier:  Biolegend 103627
Encompass_Preferred
APC/Fire™ 750 anti-mouse CD49d [R1-2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC1889100