Learn More
Biolegend APC/Fire™ 750 Rat IgG2b, κ Isotype Ctrl
Supplier:  Biolegend 400669
Encompass_Preferred
APC/Fire™ 750 Rat IgG2b, κ Isotype Ctrl [RTK4530]; Isotype: Rat IgG2b, κ; Apps: FC, ICFC; Size: 25 ug
Catalog No. NC1225713