Learn More
Biolegend APC/Fire™ 810 anti-human KLRG1 (MAFA)
Supplier:  Biolegend 367732
Encompass_Preferred
APC/Fire™ 810 anti-human KLRG1 (MAFA) [SA231A2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-213-1965