Learn More
Biolegend APC/Fire™ 810 anti-human CD14
Supplier:  Biolegend 367156
Encompass_Preferred
APC/Fire™ 810 anti-human CD14 [63D3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-8325