Learn More
Biolegend APC/Fire™ 810 Mouse IgG1, κ isotype Ctrl
Supplier:  Biolegend 402409
Encompass_Preferred
APC/Fire™ 810 Mouse IgG1, κ isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse ; Apps: FC; Size: 25 tests
Catalog No. 50-207-9684