Learn More
Biolegend APC/Fire™ 750 anti-human CD8a
Supplier:  Biolegend 300931
Encompass_Preferred
APC/Fire™ 750 anti-human CD8a [HIT8a]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Marmoset; Apps: FC; Size: 25 tests
Catalog No. 50-207-2721