Learn More
Biolegend APC/Fire™ 750 anti-human CD161
Supplier:  Biolegend 339944
Encompass_Preferred
APC/Fire™ 750 anti-human CD161 [HP-3G10]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-207-6424