Learn More
Biolegend APC/Fire™ 750 anti-human CD45RO
Supplier:  Biolegend 304250
Encompass_Preferred
APC/Fire™ 750 anti-human CD45RO [UCHL1]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Chimpanzee, Cynomolgus, Common Marmoset; Apps: FC; Size: 100 tests
Catalog No. 50-207-3078