Learn More
Biolegend APC/Fire™ 750 anti-human CD68
Supplier:  Biolegend 333824
Encompass_Preferred
APC/Fire™ 750 anti-human CD68 [Y1/82A]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. 50-207-6136