Learn More
Biolegend APC/Fire™ 750 anti-human CD7
Catalog No. 502076561
Encompass_Preferred
APC/Fire™ 750 anti-human CD7 [CD7-6B7]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-6561 Supplier Biolegend Supplier No. 343121
APC/Fire™ 750 anti-human CD7 [CD7-6B7]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests