Learn More
Biolegend APC/Fire™ 750 anti-human CD95 (Fas)
Supplier:  Biolegend 305637
Encompass_Preferred
APC/Fire™ 750 anti-human CD95 (Fas) [DX2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Chimpanzee, Common Marmoset, Cotton-topped Tamarin, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey; Apps: FC; Size: 25 tests
Catalog No. 50-207-3231