Learn More
Biolegend APC/Fire™ 750 anti-human/mouse integrin β7
Supplier:  Biolegend 321223
Encompass_Preferred
APC/Fire™ 750 anti-human/mouse integrin β7 [FIB504]; Isotype: Rat IgG2a, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Mouse, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-207-3864