Learn More
Biolegend APC/Fire™ 750 anti-human TCR Vγ9
Supplier:  Biolegend 331315
Encompass_Preferred
APC/Fire™ 750 anti-human TCR Vγ9 [B3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cynomolgus; Apps: FC; Size: 25 tests
Catalog No. 50-207-6005