Learn More
Biolegend APC/Fire™ 750 anti-human β2-microglobulin
Supplier:  Biolegend 395709
Encompass_Preferred
APC/Fire™ 750 anti-human β2-microglobulin [A17082A]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-9194