Learn More
Biolegend APC/Fire™ 750 anti-mouse CD115 (CSF-1R)
Supplier:  Biolegend 135536
Encompass_Preferred
APC/Fire™ 750 anti-mouse CD115 (CSF-1R) [AFS98]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-1128