Learn More
Biolegend APC/Fire™ 750 anti-mouse CD3ε
Supplier:  Biolegend 155614
Encompass_Preferred
APC/Fire™ 750 anti-mouse CD3ε [KT3.1.1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-2231