Learn More
Biolegend APC/Fire™ 750 anti-mouse CD90.2
Supplier:  Biolegend 105347
Encompass_Preferred
APC/Fire™ 750 anti-mouse CD90.2 [30-H12]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-206-9878