Learn More
Biolegend APC/Fire™ 750 anti-mouse Ly-6G/Ly-6C (Gr-1)
Supplier:  Biolegend 108455
Encompass_Preferred
APC/Fire™ 750 anti-mouse Ly-6G/Ly-6C (Gr-1) [RB6-8C5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0036