Learn More
Biolegend APC/Fire™ 750 anti-mouse IgG2a
Supplier:  Biolegend 407123
Encompass_Preferred
APC/Fire™ 750 anti-mouse IgG2a [RMG2a-62]; Isotype: Rat IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-9807