Learn More
Biolegend APC/FireTM 750 anti-human CD13
Catalog No. 502722515
Encompass_Preferred
APC/Fire™ 750 anti-human CD137 (4-1BB) [S18014C]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2515 Supplier Biolegend Supplier No. 300811
APC/Fire™ 750 anti-human CD137 (4-1BB) [S18014C]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests