Learn More
Biolegend APC/FireTM 750 anti-human CD14
Catalog No. 502722103
Encompass_Preferred
APC/Fire™ 750 anti-human CD14 [63D3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2103 Supplier Biolegend Supplier No. 367119
Add to Cart
Add to Cart
APC/Fire™ 750 anti-human CD14 [63D3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests