Learn More
Biolegend APC/Fire™ 750 anti-human CD301 (CLEC10A) [H037G3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 502723784
Encompass_Preferred
APC/Fire™ 750 anti-human CD301 (CLEC10A) [H037G3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-3784 Supplier Biolegend Supplier No. 354710
Add to Cart
Add to Cart
APC/Fire™ 750 anti-human CD301 (CLEC10A) [H037G3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests