Learn More
Biolegend APC/FireTM 750 anti-human CD30
Supplier:  Biolegend 354236
Encompass_Preferred
APC/Fire™ 750 anti-human CD303 (BDCA-2) [201A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-2187