Learn More
Biolegend APC/FireTM 750 anti-human MERT
Catalog No. 502722100
Encompass_Preferred
APC/Fire™ 750 anti-human MERTK [590H11G1E3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-2100 Supplier Biolegend Supplier No. 367614
APC/Fire™ 750 anti-human MERTK [590H11G1E3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests