Learn More
Biolegend APC/FireTM 750 anti-mouse CD11
Catalog No. 503108047
Encompass_Preferred
APC/Fire 750 anti-mouse CD117 (c-kit) [S18020H] Isotype Rat IgG2a Reactivity Mouse Apps FC Size 25 g
Catalog No. 50-310-8047 Supplier Biolegend Supplier No. 161609
APC/Fire 750 anti-mouse CD117 (c-kit) [S18020H] Isotype Rat IgG2a Reactivity Mouse Apps FC Size 25 g